4-Chloro-6-methylpicolinic acid hydrochloride 95%

Supplier: Ambeed
Warning

A168916-100MG A168916-250MG A168916-1G
103806-330EA 271 USD
103806-330 77151-180 77151-182 77300-952
4-Chloro-6-methylpicolinic acid hydrochloride 95%
4-Chloro-6-methylpicolinic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₇Cl₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD29044872
CAS Number: 1841081-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR