4-Chloro-6-methylnicotinic acid 98%

Supplier: Ambeed
Warning

A205076-100MG A205076-10G A205076-250MG A205076-1G A205076-25G A205076-5G
103817-590EA 16 USD
103817-590 103848-254 76873-546 76873-548 76873-550 77086-452 77086-454 77086-456
4-Chloro-6-methylnicotinic acid 98%
4-Chloro-6-methylnicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.581 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11520850
CAS Number: 1060805-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR