4-Chloro-6-methyl-7H-pyrrolo[2,3-d]pyrimidine 97%

Supplier: Ambeed
Warning

A166450-100MG A166450-250MG A166450-1G
103806-068EA 153 USD
103806-068 103844-330 76784-064 76784-066 77320-580
4-Chloro-6-methyl-7H-pyrrolo[2,3-d]pyrimidine 97%
4-Chloro-6-methyl-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₆ClN₃
MW: 167.6000 g/mol
Boiling Pt: 329.2 °C (760 mm Hg)
Melting Pt: 205 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09033903
CAS Number: 35808-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR