4-Chloro-6-methyl-2-(methylthio)pyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 4-chloro-6-methyl-2-(methylsulphanyl)pyrimidine, 4-Chloro-6-methyl-2-methylthiopyrimidine

A246344-1G A246344-25G A246344-5G
103811-704EA 18 USD
103811-704 103851-534 77090-824 77090-826 77090-828
4-Chloro-6-methyl-2-(methylthio)pyrimidine 98%
4-Chloro-6-methyl-2-(methylthio)pyrimidine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂S
MW: 174.65 g/mol
Boiling Pt: 266.2±20.0 °C (760 mm Hg)
Flash Pt: 114.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00023233
CAS Number: 17119-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR