4-Chloro-6-methyl-1H-indole 98%

Supplier: Ambeed
Warning

A233973-100MG A233973-250MG A233973-50MG
103810-920EA 204 USD
103810-920 103850-680 77013-606 77013-608 77013-610
4-Chloro-6-methyl-1H-indole 98%
4-Chloro-6-methyl-1H-indole

Precaución: for Research Use Only

Formula: C₉H₈ClN
MW: 165.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781504
CAS Number: 885521-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR