4-Chloro-6-methoxyquinolin-7-ol 95%

Supplier: Ambeed
Warning

A588254-1G A588254-100MG A588254-10G A588254-250MG A588254-25G A588254-5G
103828-908EA 36 USD
103828-908 103840-052 76844-196 77035-052 77035-054 77035-056 77307-898 77307-900
4-Chloro-6-methoxyquinolin-7-ol 95%
4-Chloro-6-methoxyquinolin-7-ol

Precaución: for Research Use Only

Formula: C₁₀H₈ClNO₂
MW: 209.6290 g/mol
Boiling Pt: 353 °C (760 mm Hg)
Flash Pt: 168 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13191788
CAS Number: 205448-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR