4-Chloro-6-methoxy-2-methylpyrimidine 95%

Supplier: Ambeed
Warning

A250599-100MG A250599-10G A250599-250MG A250599-25G A250599-1G A250599-5G
103811-932EA 16 USD
103811-932 103851-770 77031-504 77031-506 77031-508 77031-510 77304-532 77304-534
4-Chloro-6-methoxy-2-methylpyrimidine 95%
4-Chloro-6-methoxy-2-methylpyrimidine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O
Flash Pt: 87.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16768933
CAS Number: 89466-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR