4-Chloro-6-methoxy-2-(methylsulfonyl)pyrimidine 97%

Supplier: Ambeed
Warning

A155652-1G A155652-5G A155652-250MG
103804-786EA 45 USD
103804-786 103842-988 77300-144 77300-146 77671-200
4-Chloro-6-methoxy-2-(methylsulfonyl)pyrimidine 97%
4-Chloro-6-methoxy-2-(methylsulfonyl)pyrimidine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O₃S
MW: 222.65 g/mol
Boiling Pt: 199.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD10574963
CAS Number: 89466-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR