4-Chloro-6-iodoquinazoline 95%

Supplier: Ambeed
Warning

A230716-1G A230716-25G A230716-100G A230716-10G A230716-5G
103810-724EA 6 USD
103810-724 103850-466 77089-336 77089-338 77288-360 77288-362 77303-884
4-Chloro-6-iodoquinazoline 95%
4-Chloro-6-iodoquinazoline

Precaución: for Research Use Only

Formula: C₈H₄ClIN₂
MW: 290.4900 g/mol
Boiling Pt: 363.2 °C (760 mm Hg)
Melting Pt: 175…177 °C
Flash Pt: 173.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01862193
CAS Number: 98556-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR