4-Chloro-6-hydroxyquinoline 97%

Supplier: Ambeed
Warning

A352245-100MG A352245-250MG A352245-1G
103826-062EA 93 USD
103826-062 103837-062 77033-112 77033-114 77306-628
4-Chloro-6-hydroxyquinoline 97%
4-Chloro-6-hydroxyquinoline

Precaución: for Research Use Only

Formula: C₉H₆ClNO
Boiling Pt: 304.9 °C (760 mm Hg)
Flash Pt: 138.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00456102
CAS Number: 148018-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR