4-Chloro-6-fluoro-2-methylquinazoline 98%

Supplier: Ambeed
Warning

A386004-1G A386004-250MG
103826-792EA 66 USD
103826-792 103837-832 76841-184 77033-480
4-Chloro-6-fluoro-2-methylquinazoline 98%
4-Chloro-6-fluoro-2-methylquinazoline

Precaución: for Research Use Only

Formula: C₉H₆ClFN₂
Boiling Pt: 216 °C (760 mm Hg)
Flash Pt: 84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11047031
CAS Number: 1044768-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR