4-Chloro-6-ethyl-5-fluoropyrimidine 98%

Supplier: Ambeed
Warning

A102502-100G A102502-10G A102502-25G A102502-5G A102502-500G
103822-072EA 6 USD
103822-072 77282-934 77282-936 77282-938 77282-940 77747-764
4-Chloro-6-ethyl-5-fluoropyrimidine 98%
4-Chloro-6-ethyl-5-fluoropyrimidine

Precaución: for Research Use Only

Formula: C₆H₆ClFN₂
Boiling Pt: 210.9 °C (760 mm Hg)
Flash Pt: 81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07782087
CAS Number: 137234-74-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR