4-Chloro-6,7-dimethoxyquinazoline 98%

Supplier: Ambeed
Warning

A176062-1G A176062-25G A176062-5G A176062-100G A176062-10G
103807-112EA 6 USD
103807-112 103845-428 77082-778 77082-780 77082-782 77273-884 77273-886
4-Chloro-6,7-dimethoxyquinazoline 98%
4-Chloro-6,7-dimethoxyquinazoline

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂O₂
MW: 224.65 g/mol
Boiling Pt: 345.5 °C (760 mm Hg)
Melting Pt: 185 °C
Flash Pt: 162.8 °C
Storage Temperature: Ambient
MDL Number: MFCD01570172
CAS Number: 13790-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR