4-Chloro-6,7-dimethoxy-2-methylquinazoline 97%

Supplier: Ambeed
Warning

A124197-250MG A124197-1G
103800-502EA 40 USD
103800-502 77026-682 77312-634
4-Chloro-6,7-dimethoxy-2-methylquinazoline 97%
4-Chloro-6,7-dimethoxy-2-methylquinazoline

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClN₂O₂
MW: 238.67 g/mol
Boiling Pt: 286.8±40.0 °C (760 mm Hg)
Flash Pt: 127.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11046864
CAS Number: 50377-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR