4-Chloro-6,7-dihydro-5H-cyclopenta[b]pyridin-7-ol 98%

Supplier: Ambeed
Warning

A173955-100MG A173955-1G A173955-5G A173955-250MG
103806-892EA 95 USD
103806-892 103845-190 77029-208 77301-300 77301-302 77593-090
4-Chloro-6,7-dihydro-5H-cyclopenta[b]pyridin-7-ol 98%
4-Chloro-6,7-dihydro-5H-cyclopenta[b]pyridin-7-ol

Precaución: for Research Use Only

Formula: C₈H₈ClNO
MW: 169.6100 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13152137
CAS Number: 126053-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR