4-Chloro-6,7-bis(2-methoxyethoxy)quinazoline 98%

Supplier: Ambeed
Warning

A152496-10G A152496-25G A152496-5G A152496-100G
103804-400EA 8 USD
103804-400 103842-570 76997-398 77299-956 77318-190 77671-136
4-Chloro-6,7-bis(2-methoxyethoxy)quinazoline 98%
4-Chloro-6,7-bis(2-methoxyethoxy)quinazoline

Precaución: for Research Use Only

Formula: C₁₄H₁₇ClN₂O₄
Boiling Pt: 446.7 °C (760 mm Hg)
Melting Pt: 105…107 °C
Flash Pt: 224 °C
Storage Temperature: Ambient
MDL Number: MFCD09751265
CAS Number: 183322-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR