2-Amino-4-chloro-5-nitropyridine 96%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-5-nitropyridin-2-amine

A307795-1G A307795-25G
103814-524EA 13 USD
103814-524 103835-700 76838-446 77167-064
2-Amino-4-chloro-5-nitropyridine 96%
2-Amino-4-chloro-5-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₄ClN₃O₂
MW: 173.557 g/mol
Boiling Pt: 203.1 °C (760 mm Hg)
Melting Pt: 86.8…87.0 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04973545
CAS Number: 24484-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR