4-Chloro-5-nitropyridin-2(1H)-one 97%

Supplier: Ambeed
Warning

A106148-10G A106148-250MG A106148-25G A106148-1G A106148-5G
103797-004EA 47 USD
103797-004 103822-758 76780-824 76780-826 76780-828 77309-616 77309-618
4-Chloro-5-nitropyridin-2(1H)-one 97%
4-Chloro-5-nitropyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₅H₃ClN₂O₃
Storage Temperature: Freezer
MDL Number: MFCD11046303
CAS Number: 850663-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR