4-Chloro-5-methylpyridin-2-amine 97%
Supplier: Ambeed
|
Warning
|
A210788-250MG
A210788-1G
103818-042EA
149
USD
103818-042
76873-668
77087-120
4-Chloro-5-methylpyridin-2-amine 97%
4-Chloro-5-methylpyridin-2-amine
Precaución: for Research Use Only
Formula:
C₆H₇ClN₂ MW: 142.59 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11100639 CAS Number: 1033203-31-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...