4-Chloro-5-methyl-7H-pyrrolo[2,3-d]pyrimidine 97%

Supplier: Ambeed
Warning

A175948-1G A175948-5G A175948-100MG A175948-250MG
103807-100EA 81 USD
103807-100 103845-412 77082-760 77082-762 77138-538 77138-540
4-Chloro-5-methyl-7H-pyrrolo[2,3-d]pyrimidine 97%
4-Chloro-5-methyl-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₆ClN₃
Boiling Pt: 279.9 °C (760 mm Hg)
Flash Pt: 123 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09264586
CAS Number: 1618-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR