4-Chloro-5-methoxy-2-(methylsulfanyl)pyrimidine 95%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-5-methoxy-2-(methylthio)pyrimidine

A111461-100MG A111461-1G A111461-250MG
103797-836EA 68 USD
103797-836 77284-116 77284-118 77284-120
4-Chloro-5-methoxy-2-(methylsulfanyl)pyrimidine 95%
4-Chloro-5-methoxy-2-(methylsulfanyl)pyrimidine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂OS
MW: 190.65 g/mol
Boiling Pt: 285.5 °C (760 mm Hg)
Melting Pt: 73…77 °C
Flash Pt: 126.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00223689
CAS Number: 87026-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR