4-Chloro-5-iodopyridin-2-amine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4-chloro-5-iodopyridine

A199039-250MG A199039-1G
103817-074EA 11 USD
103817-074 103847-698 76986-374 77085-736
4-Chloro-5-iodopyridin-2-amine 98%
4-Chloro-5-iodopyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄ClIN₂
Boiling Pt: 326.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11977433
CAS Number: 670253-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR