6-Chloro-7-iodo-7-deazapurine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine, 6-Chloro-7-iodo-deazapurine
A167986-100G
A167986-10G
A167986-500G
A167986-5G
A167986-1G
A167986-25G
103806-236EA
6
USD
103806-236
103844-494
76998-102
76998-104
76998-106
77081-532
77300-912
77300-914
6-Chloro-7-iodo-7-deazapurine 98%
6-Chloro-7-iodo-7-deazapurine
Precaución: for Research Use Only
Formula:
C₆H₃ClIN₃ MW: 279.47 g/mol Boiling Pt: 241.0±50.0 °C (760 mm Hg) Melting Pt: 179…183 °C Flash Pt: 99.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD09263258 CAS Number: 123148-78-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...