4-Chloro-5H-pyrrolo[3,2-d]pyrimidine 98%
Supplier: Ambeed
|
Warning
|
A234764-1G
A234764-25G
A234764-5G
A234764-10G
A234764-250MG
A234764-100G
103810-974EA
8
USD
103810-974
103850-730
77089-728
77089-730
77089-732
77165-654
77165-656
77892-401
4-Chloro-5H-pyrrolo[3,2-d]pyrimidine 98%
4-Chloro-5H-pyrrolo[3,2-d]pyrimidine
Precaución: for Research Use Only
Formula:
C₆H₄ClN₃ Melting Pt: 325.9 °C Flash Pt: 180.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06658411 CAS Number: 84905-80-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...