4-Chloro-5-fluoropyrimidine 95%

Supplier: Ambeed
Warning

A200204-1G A200204-25G A200204-5G
103817-174EA 112 USD
103817-174 103847-802 77302-660 77302-662 77302-664
4-Chloro-5-fluoropyrimidine 95%
4-Chloro-5-fluoropyrimidine

Precaución: for Research Use Only

Formula: C₄H₂ClFN₂
Boiling Pt: 181.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09750157
CAS Number: 347418-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR