4-Chloro-5-fluoro-2-nitroaniline 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-5-chloro-4-fluoronitrobenzene
A168899-1G
A168899-5G
103806-328EA
20
USD
103806-328
103844-584
77081-654
77081-656
4-Chloro-5-fluoro-2-nitroaniline 97%
4-Chloro-5-fluoro-2-nitroaniline
Precaución: for Research Use Only
Formula:
C₆H₄ClFN₂O₂ MW: 190.56 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12828238 CAS Number: 428871-64-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...