4-Chloro-5-fluoro-2-methylpyrimidine 96%

Supplier: Ambeed
Warning

A209096-1G A209096-5G
103817-916EA 27 USD
103817-916 103848-602 77086-940 77086-942
4-Chloro-5-fluoro-2-methylpyrimidine 96%
4-Chloro-5-fluoro-2-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₄ClFN₂
MW: 146.5501 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09834964
CAS Number: 898044-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR