4-Chloro-5-fluoro-2-hydroxybenzaldehyde 97%

Supplier: Ambeed
Warning

A219416-1G A219416-100MG A219416-250MG
103818-794EA 131 USD
103818-794 103849-672 77088-230 77165-162 77165-164
4-Chloro-5-fluoro-2-hydroxybenzaldehyde 97%
4-Chloro-5-fluoro-2-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClFO₂
MW: 174.557 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20278246
CAS Number: 1205551-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR