4-Chloro-3-nitropyridine 95%
Supplier: Ambeed
|
Danger
|
A183366-5G
A183366-10G
A183366-25G
103815-634EA
6
USD
103815-634
103846-196
77083-800
77567-680
77567-682
4-Chloro-3-nitropyridine 95%
4-Chloro-3-nitropyridine
Precaución: For research use only.
Formula:
C₅H₃ClN₂O₂ MW: 158.54 g/mol Boiling Pt: 246.5 °C (760 mm Hg) Melting Pt: 32 °C Storage Temperature: Freezer |
MDL Number:
MFCD02683025 CAS Number: 13091-23-1 UN: 2923 ADR: 8,6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...