2-Amino-4-chloro-3-nitropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-3-nitropyridin-2-amine

A605328-1G A605328-25G A605328-5G A605328-10G A605328-250MG
103840-138EA 41 USD
103840-138 76844-300 76844-302 76844-304 77254-280 77254-282
2-Amino-4-chloro-3-nitropyridine 98%
2-Amino-4-chloro-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₄ClN₃O₂
MW: 173.56 g/mol
Boiling Pt: 329 °C (760 mm Hg)
Melting Pt: 174…176 °C
Flash Pt: 152.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04116097
CAS Number: 6980-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR