4-Chloro-3-nitrophenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Chloro-3-nitrobenzeneboronic acid
A796326-1G
A796326-25G
A796326-5G
A796326-250MG
103841-042EA
45
USD
103841-042
76845-292
76845-294
76845-296
77105-558
4-Chloro-3-nitrophenylboronic acid 98%
4-Chloro-3-nitrophenylboronic acid
Precaución: for Research Use Only
Formula:
C₆H₅BClNO₄ Boiling Pt: 385.4 °C (760 mm Hg) Melting Pt: 270…278 °C Flash Pt: 186.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02258950 CAS Number: 151169-67-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...