4-Chloro-3-nitrophenol 98%

Supplier: Ambeed
Danger

A433287-1G A433287-25G A433287-5G A433287-100G
103827-548EA 5 USD
103827-548 103838-614 76842-318 76842-320 76842-322 77102-312
4-Chloro-3-nitrophenol 98%
4-Chloro-3-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₄ClNO₃
Boiling Pt: 308.8 °C (760 mm Hg)
Melting Pt: 125…129 °C (760 mm Hg)
Flash Pt: 140.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00043546
CAS Number: 610-78-6
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR