4-Chloro-3-nitrobenzenesulfonyl chloride 96%

Supplier: Ambeed
Danger

Synonyms: 4-Chloro-3-nitrobenzenesulphonyl Chloride, 4-Chloro-3-nitrobenzene-1-sulphonyl chloride

A419747-1G A419747-25G A419747-5G
103827-372EA 7 USD
103827-372 103838-422 76842-040 76842-042 76842-044
4-Chloro-3-nitrobenzenesulfonyl chloride 96%
4-Chloro-3-nitrobenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO₄S
MW: 256.07 g/mol
Melting Pt: 59…60 °C
Flash Pt: 169.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00007440
CAS Number: 97-08-5
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR