4-Chloro-3-methylphenylboronic acid 98%

Supplier: Ambeed
Warning

A203992-1G A203992-10G A203992-250MG A203992-25G
103817-514EA 6 USD
103817-514 103848-176 77086-324 77192-412 77192-414 77302-892
4-Chloro-3-methylphenylboronic acid 98%
4-Chloro-3-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BClO₂
Boiling Pt: 330.1 °C (760 mm Hg)
Flash Pt: 153.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683104
CAS Number: 161950-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR