4-Chloro-3-methylbenzaldehyde 97%

Supplier: Ambeed
Warning

A407114-1G A407114-250MG
103827-152EA 48 USD
103827-152 103838-192 76841-686 77890-837
4-Chloro-3-methylbenzaldehyde 97%
4-Chloro-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇ClO
Boiling Pt: 234 °C (760 mm Hg)
Melting Pt: 27…28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02261736
CAS Number: 101349-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR