4-Chloro-3-methyl-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Ambeed
Warning

A161051-250MG A161051-1G
103805-446EA 53 USD
103805-446 103843-684 77028-646 77319-650
4-Chloro-3-methyl-1H-pyrrolo[2,3-b]pyridine 95%
4-Chloro-3-methyl-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₈H₇ClN₂
MW: 166.6100 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15529289
CAS Number: 688782-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR