4-Chloro-3-methoxypyridin-2-amine 98%

Supplier: Ambeed
Warning

A171235-100MG A171235-250MG
103806-606EA 213 USD
103806-606 77138-420 77138-422
4-Chloro-3-methoxypyridin-2-amine 98%
4-Chloro-3-methoxypyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD18415420
CAS Number: 1261452-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR