4-Chloro-3-methoxyphenol 95%

Supplier: Ambeed
Warning

A885993-1G A885993-100MG A885993-250MG
103841-326EA 352 USD
103841-326 76845-594 77159-698 77159-700
4-Chloro-3-methoxyphenol 95%
4-Chloro-3-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇ClO₂
MW: 158.582 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16998742
CAS Number: 18113-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR