2-Amino-4-chloro-3-iodopyridine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-3-iodopyridin-2-amine

A438521-1G A438521-25G A438521-5G A438521-250MG
103827-634EA 28 USD
103827-634 103838-698 76842-440 76842-442 76842-444 76990-584
2-Amino-4-chloro-3-iodopyridine 98%
2-Amino-4-chloro-3-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₄ClIN₂
MW: 254.46 g/mol
Boiling Pt: 319.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD08741521
CAS Number: 417721-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR