4-Chloro-3-iodo-1H-pyrazolo[4,3-c]pyridine 96%
Supplier: Ambeed
|
Danger
|
A502480-5G
A502480-100MG
A502480-250MG
103828-302EA
22
USD
103828-302
103839-392
76843-360
76979-872
77673-314
4-Chloro-3-iodo-1H-pyrazolo[4,3-c]pyridine 96%
4-Chloro-3-iodo-1H-pyrazolo[4,3-c]pyridine
Precaución: For research use only
Formula:
C₆H₃ClIN₃ MW: 279.4700 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD16877655 CAS Number: 1186647-69-7 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...