4-Chloro-3-hydroxybenzoic acid 98%

Supplier: Ambeed
Warning

A203440-1G A203440-5G A203440-100G A203440-10G A203440-250MG A203440-25G
103817-462EA 7 USD
103817-462 103848-116 77086-252 77086-254 77139-398 77139-400 77139-402 77302-850
4-Chloro-3-hydroxybenzoic acid 98%
4-Chloro-3-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅ClO₃
Boiling Pt: 457.8 °C (760 mm Hg)
Melting Pt: 230…238 °C
Flash Pt: 162.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00153892
CAS Number: 34113-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR