4-Chloro-3-hydroxybenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A116134-10G
A116134-250MG
A116134-25G
A116134-5G
A116134-1G
103798-678EA
32
USD
103798-678
103824-542
76810-204
76810-206
76810-208
77297-424
77311-360
4-Chloro-3-hydroxybenzaldehyde 98%
4-Chloro-3-hydroxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₅ClO₂ MW: 156.5700 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD16999770 CAS Number: 56962-12-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...