4-Chloro-3-fluoropyridine 98% (by GC)

Supplier: Ambeed
Danger

A306455-1G A306455-25G A306455-10G
103814-464EA 7 USD
103814-464 103835-630 76838-334 76838-336 77290-104
4-Chloro-3-fluoropyridine 98% (by GC)
4-Chloro-3-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₃ClFN
MW: 131.54 g/mol
Boiling Pt: 138.9 °C (760 mm Hg)
Flash Pt: 28 °C
Storage Temperature: Freezer
MDL Number: MFCD03453233
CAS Number: 2546-56-7
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR