4-Chloro-3-fluorophenol 98%
Supplier: Ambeed
|
Warning
|
A164485-100G
A164485-10G
A164485-500G
A164485-1G
A164485-25G
A164485-5G
103805-868EA
9
USD
103805-868
103844-128
77286-564
77286-566
77286-568
77320-300
77320-302
77320-304
4-Chloro-3-fluorophenol 98%
4-Chloro-3-fluorophenol
Precaución: for Research Use Only
Formula:
C₆H₄ClFO MW: 146.55 g/mol Boiling Pt: 163.3 °C (760 mm Hg) Melting Pt: 54…56 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042583 CAS Number: 348-60-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...