1-Chloro-2-fluoro-4-iodobenzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Chloro-3-fluoroiodobenzene
A192226-25G
A192226-5G
A192226-10G
A192226-1G
A192226-100G
103816-424EA
9
USD
103816-424
103847-030
77084-902
77084-904
77097-150
77302-284
77685-466
1-Chloro-2-fluoro-4-iodobenzene 98%
1-Chloro-2-fluoro-4-iodobenzene
Precaución: for Research Use Only
Formula:
C₆H₃ClFI Boiling Pt: 221.7 °C (760 mm Hg) Flash Pt: 87.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042580 CAS Number: 202982-67-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...