4-Chloro-3-fluorobenzoic acid 97%

Supplier: Ambeed
Warning

A322344-100G A322344-10G A322344-1G A322344-25G A322344-5G
103825-302EA 5 USD
103825-302 103836-234 77141-832 77141-834 77306-258 77306-260 77306-262
4-Chloro-3-fluorobenzoic acid 97%
4-Chloro-3-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄ClFO₂
MW: 174.56 g/mol
Boiling Pt: 290.9±20.0 °C (760 mm Hg)
Melting Pt: 190…196 °C
Flash Pt: 129.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00143290
CAS Number: 403-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR