4-Chloro-3-fluoro-2-iodoaniline 97%

Supplier: Ambeed
Warning

A185056-250MG A185056-1G A185056-5G
103815-810EA 55 USD
103815-810 103846-376 77012-090 77084-010 77084-012
4-Chloro-3-fluoro-2-iodoaniline 97%
4-Chloro-3-fluoro-2-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₄ClFIN
Boiling Pt: 299.7 °C (760 mm Hg)
Melting Pt: 80.5…81.0 °C
Flash Pt: 135.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18250658
CAS Number: 1018450-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR