4-Chloro-3-(trifluoromethyl)phenol 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-hydroxybenzotrifluoride

A589092-25G A589092-5G A589092-100G A589092-10G A589092-1G
103828-916EA 6 USD
103828-916 103840-060 76844-204 76844-206 77117-218 77117-220 77307-904
4-Chloro-3-(trifluoromethyl)phenol 97%
4-Chloro-3-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₄ClF₃O
MW: 196.56 g/mol
Boiling Pt: 232 °C (760 mm Hg)
Melting Pt: 44…46 °C
Storage Temperature: Ambient
MDL Number: MFCD00042525
CAS Number: 6294-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR