4-Chloro-3-(trifluoromethoxy)benzoic acid 98%
Supplier: Ambeed
|
Warning
|
A196079-250MG
A196079-1G
A196079-5G
103816-810EA
42
USD
103816-810
103847-424
76785-102
77085-376
77302-500
4-Chloro-3-(trifluoromethoxy)benzoic acid 98%
4-Chloro-3-(trifluoromethoxy)benzoic acid
Precaución: for Research Use Only
Formula:
C₈H₄ClF₃O₃ MW: 240.57 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06660238 CAS Number: 886500-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...