4-Chloro-2-nitropyridine 97%
Supplier: Ambeed
|
Warning
|
A325381-1G
A325381-25G
A325381-5G
A325381-100G
A325381-10G
A325381-250MG
103825-400EA
17
USD
103825-400
103836-344
76839-276
76839-278
76839-280
77290-346
77290-348
77290-350
4-Chloro-2-nitropyridine 97%
4-Chloro-2-nitropyridine
Precaución: for Research Use Only
Formula:
C₅H₃ClN₂O₂ Boiling Pt: 267.6 °C (760 mm Hg) Flash Pt: 115.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04114167 CAS Number: 65370-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...